Assay Details
Gene Symbol: TANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277871.1 | NP_001264800.1 | 5 - 6 | 629 | 101 | |
XM_015289646.2 | XP_015145132.1 | 5 - 6 | 708 | 101 | ||
XM_025152196.1 | XP_025007964.1 | 5 - 6 | 575 | 101 | ||
XM_025152197.1 | XP_025007965.1 | 6 - 7 | 1595 | 101 | ||
GenBank mRNA | BX929866.2 | - | 5 - 6 | 617 | 101 | |
CR354363.1 | - | 5 - 6 | 629 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
