Assay Details
Gene Symbol: CYBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415607.6 | XP_415607.1 | 2 - 3 | 336 | 67 | |
GenBank mRNA | BX933442.2 | - | 2 - 3 | 209 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
