Assay Details
Gene Symbol: CYBC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415607.6 | XP_415607.1 | 6 - 7 | 699 | 101 | |
GenBank mRNA | BX933442.2 | - | 6 - 7 | 572 | 101 | |
BX950273.2 | - | 3 - 4 | 265 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
