Assay Details
Gene Symbol: RPLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_424134.6 | XP_424134.1 | 1 - 2 | 72 | 59 | |
GenBank mRNA | BX935520.2 | - | 1 - 2 | 55 | 59 | |
BX935793.2 | - | 1 - 2 | 49 | 59 | ||
CR406227.1 | - | 1 - 2 | 89 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_424134.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
