Assay Details
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277590.1 | NP_001264519.1 | 4 - 5 | 386 | 62 | |
GenBank mRNA | BM440029.1 | - | 4 - 5 | 381 | 62 | |
BU380556.1 | - | 3 - 4 | 371 | 62 | ||
BX540414.3 | - | 4 - 5 | 393 | 62 | ||
BX950422.2 | - | 4 - 5 | 379 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
