Assay Details
Gene Symbol: CYLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015292398.2 | XP_015147884.2 | 7 - 8 | 1759 | 134 | |
GenBank mRNA | CR386539.1 | - | 4 - 5 | 657 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
