Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025155393.1 | XP_025011161.1 | 3 | 4972 | 81 | |
XM_429544.6 | XP_429544.6 | 3 | 4952 | 81 | ||
GenBank mRNA | CR387739.1 | - | 1 | 749 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
