Assay Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004940220.3 | XP_004940277.3 | 4 - 5 | 3137 | 80 | |
XM_004940221.3 | XP_004940278.3 | 4 - 5 | 3164 | 80 | ||
GenBank mRNA | BX929377.1 | - | 4 - 5 | 619 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
