Assay Details
Gene Symbol: LOC425607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025146508.1 | XP_025002276.1 | 1254 | 83 | ||
GenBank mRNA | BX932672.1 | - | 51 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
