Assay Details
Gene Symbol: ZGPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004947125.2 | XP_004947182.1 | 1 - 2 | 927 | 67 | |
GenBank mRNA | BX934290.1 | - | 1 - 2 | 484 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
