Assay Details
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288444.2 | XP_015143930.1 | 4 - 5 | 400 | 84 | |
XM_421646.6 | XP_421646.1 | 4 - 5 | 306 | 84 | ||
GenBank mRNA | BX935521.1 | - | 5 - 6 | 308 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
