Assay Details
Gene Symbol: SRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205457.3 | NP_990788.3 | 3 - 4 | 415 | 72 | |
XM_015296348.2 | XP_015151834.2 | 4 - 5 | 754 | 72 | ||
GenBank mRNA | M57290.1 | - | 4 - 5 | 2482 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
