Assay Details
Gene Symbol: SRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205457.3 | NP_990788.3 | 4 - 5 | 516 | 118 | |
XM_015296346.2 | XP_015151832.2 | 6 - 7 | 681 | 118 | ||
XM_015296347.2 | XP_015151833.2 | 5 - 6 | 666 | 118 | ||
XM_015296348.2 | XP_015151834.2 | 5 - 6 | 855 | 118 | ||
GenBank mRNA | M57290.1 | - | 5 - 6 | 2583 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
