Assay Details
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277696.1 | NP_001264625.1 | 5 - 6 | 698 | 67 | |
XM_015293646.2 | XP_015149132.1 | 13 - 14 | 2483 | 67 | ||
XM_025154824.1 | XP_025010592.1 | 13 - 14 | 2479 | 67 | ||
XM_025154825.1 | XP_025010593.1 | 14 - 15 | 1637 | 67 | ||
XM_025154826.1 | XP_025010594.1 | 7 - 8 | 927 | 67 | ||
GenBank mRNA | X72806.1 | - | 735 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
