Assay Details
Gene Symbol: KDELR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159715.1 | NP_001153187.1 | 3 - 4 | 464 | 82 | |
GenBank mRNA | AJ719955.1 | - | 3 - 4 | 464 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
