Assay Details
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR406617.1 | - | 1 | 450 | 105 |
Gene Symbol: LOC776067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277919.2 | XP_015133405.2 | 11 | 6576 | 105 | |
XM_025146674.1 | XP_025002442.1 | 21 | 9189 | 105 | ||
XM_025146675.1 | XP_025002443.1 | 21 | 9189 | 105 | ||
XM_025146676.1 | XP_025002444.1 | 20 | 5893 | 105 | ||
XM_025146677.1 | XP_025002445.1 | 21 | 9186 | 105 | ||
XM_025146678.1 | XP_025002446.1 | 21 | 9189 | 105 | ||
XM_025146682.1 | XP_025002450.1 | 11 | 6576 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
