Assay Details
Gene Symbol: GPAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297404.2 | XP_015152890.1 | 2 - 3 | 951 | 72 | |
GenBank mRNA | CR524251.1 | - | 1 - 2 | 475 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
