Assay Details
Gene Symbol: MYDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006342.1 | NP_001006342.1 | 2 - 3 | 236 | 87 | |
GenBank mRNA | AJ719430.1 | - | 2 - 3 | 236 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
