Assay Details
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006345.2 | NP_001006345.1 | 4 - 5 | 447 | 111 | |
GenBank mRNA | AJ720578.1 | - | 4 - 5 | 441 | 111 | |
BU135171.1 | - | 4 - 5 | 447 | 111 | ||
CR338953.1 | - | 4 - 5 | 427 | 111 | ||
CR352384.1 | - | 3 - 4 | 417 | 111 | ||
CR353058.1 | - | 4 - 5 | 424 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
