Assay Details
Gene Symbol: FABP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006346.1 | NP_001006346.1 | 1 - 2 | 134 | 73 | |
GenBank mRNA | AJ720717.1 | - | 1 - 2 | 134 | 73 | |
BX936126.1 | - | 1 - 2 | 303 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
