Assay Details
Gene Symbol: ENPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006422.2 | NP_001006422.2 | 3 - 4 | 1178 | 136 | |
XM_015285142.2 | XP_015140628.1 | 3 - 4 | 1100 | 136 | ||
GenBank mRNA | AJ720160.1 | - | 3 - 4 | 1177 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
