Assay Details
Gene Symbol: CHMP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006428.1 | NP_001006428.1 | 6 - 7 | 545 | 72 | |
GenBank mRNA | AJ719963.1 | - | 6 - 7 | 545 | 72 | |
CR387811.1 | - | 6 - 7 | 562 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
