Assay Details
Gene Symbol: SRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006478.2 | NP_001006478.1 | 1 - 2 | 80 | 61 | |
GenBank mRNA | AJ720841.1 | - | 1 - 2 | 81 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
