Assay Details
Gene Symbol: ID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204590.2 | NP_989921.1 | 1 - 2 | 323 | 134 | |
GenBank mRNA | AY040527.1 | - | 1 - 2 | 749 | 134 | |
DR427089.1 | - | 1 - 2 | 323 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
