Assay Details
Gene Symbol: TRAIL-LIKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204591.2 | NP_989922.1 | 1 - 2 | 300 | 96 | |
GenBank mRNA | AY057941.1 | - | 1 - 2 | 304 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
