Assay Details
Gene Symbol: TRAIL-LIKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204591.2 | NP_989922.1 | 3 - 4 | 477 | 62 | |
XM_015278184.2 | XP_015133670.1 | 3 - 4 | 468 | 62 | ||
GenBank mRNA | AY057941.1 | - | 3 - 4 | 481 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
