Assay Details
Gene Symbol: OPN4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204625.3 | NP_989956.3 | 2 - 3 | 274 | 107 | |
XM_015276115.2 | XP_015131601.1 | 12 - 13 | 3100 | 107 | ||
XM_015276116.2 | XP_015131602.1 | 12 - 13 | 8064 | 107 | ||
XM_015276120.2 | XP_015131606.1 | 4 - 5 | 1153 | 107 | ||
XM_025149493.1 | XP_025005261.1 | 12 - 13 | 8138 | 107 | ||
XM_025149494.1 | XP_025005262.1 | 10 - 11 | 8168 | 107 | ||
XM_025149495.1 | XP_025005263.1 | 12 - 13 | 5276 | 107 | ||
XM_025149496.1 | XP_025005264.1 | 11 - 12 | 8142 | 107 | ||
XM_025149497.1 | XP_025005265.1 | 12 - 13 | 8129 | 107 | ||
XM_025149498.1 | XP_025005266.1 | 11 - 12 | 7954 | 107 | ||
GenBank mRNA | AY036061.1 | - | 2 - 3 | 276 | 107 | |
EU124630.1 | - | 2 - 3 | 276 | 107 | ||
EU124631.1 | - | 2 - 3 | 276 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
