Assay Details
Gene Symbol: OPN4-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204625.3 | NP_989956.3 | 6 - 7 | 918 | 80 | |
XM_015276115.2 | XP_015131601.1 | 16 - 17 | 3744 | 80 | ||
XM_015276116.2 | XP_015131602.1 | 16 - 17 | 8708 | 80 | ||
XM_015276120.2 | XP_015131606.1 | 8 - 9 | 1797 | 80 | ||
XM_025149493.1 | XP_025005261.1 | 16 - 17 | 8782 | 80 | ||
XM_025149494.1 | XP_025005262.1 | 14 - 15 | 8812 | 80 | ||
XM_025149495.1 | XP_025005263.1 | 16 - 17 | 5920 | 80 | ||
XM_025149496.1 | XP_025005264.1 | 15 - 16 | 8786 | 80 | ||
XM_025149497.1 | XP_025005265.1 | 16 - 17 | 8773 | 80 | ||
XM_025149498.1 | XP_025005266.1 | 15 - 16 | 8598 | 80 | ||
GenBank mRNA | AY036061.1 | - | 6 - 7 | 920 | 80 | |
EU124630.1 | - | 6 - 7 | 920 | 80 | ||
EU124631.1 | - | 6 - 7 | 920 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
