Assay Details
Gene Symbol: MINPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204644.1 | NP_989975.1 | 3 - 4 | 898 | 141 | |
GenBank mRNA | U59421.1 | - | 3 - 4 | 898 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
