Assay Details
Gene Symbol: SALL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204707.1 | NP_990038.1 | 2 - 3 | 3951 | 82 | |
XM_025154146.1 | XP_025009914.1 | 3 - 4 | 3642 | 82 | ||
GenBank mRNA | AF288697.1 | - | 2 - 3 | 3951 | 82 | |
CR387192.1 | - | 1 - 2 | 259 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
