Assay Details
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204721.2 | NP_990052.1 | 2 - 3 | 418 | 80 | |
XM_025149500.1 | XP_025005268.1 | 2 - 3 | 317 | 80 | ||
GenBank mRNA | AB033829.1 | - | 2 - 3 | 420 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
