Assay Details
Gene Symbol: CHRNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204812.2 | NP_990143.2 | 1 - 2 | 56 | 114 | |
XM_015280411.2 | XP_015135897.1 | 1 - 2 | 497 | 114 | ||
GenBank mRNA | X83739.2 | - | 56 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
