Assay Details
Gene Symbol: CHRNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204812.2 | NP_990143.2 | 2 - 3 | 209 | 81 | |
XM_015280411.2 | XP_015135897.1 | 2 - 3 | 650 | 81 | ||
XM_025144652.1 | XP_025000420.1 | 1 - 2 | 2082 | 81 | ||
XM_025144653.1 | XP_025000421.1 | 1 - 2 | 2078 | 81 | ||
GenBank mRNA | X83739.2 | - | 209 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
