Assay Details
Gene Symbol: CHRNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204814.1 | NP_990145.1 | 1 - 2 | 72 | 64 | |
GenBank mRNA | AJ250361.1 | - | 1 - 2 | 72 | 64 | |
BX931114.1 | - | 1 - 2 | 662 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
