Assay Details
Gene Symbol: CHRNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204814.1 | NP_990145.1 | 4 - 5 | 382 | 61 | |
GenBank mRNA | AJ250361.1 | - | 4 - 5 | 382 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
