Assay Details
Gene Symbol: CHRNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204815.2 | NP_990146.1 | 5 - 6 | 1467 | 56 | |
GenBank mRNA | AJ250360.1 | - | 5 - 6 | 1469 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
