Assay Details
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204890.1 | NP_990221.1 | 3 - 4 | 516 | 64 | |
GenBank mRNA | X60465.1 | - | 516 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
