Assay Details
Gene Symbol: RAB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205228.1 | NP_990559.1 | 4 - 5 | 272 | 128 | |
GenBank mRNA | X84220.1 | - | 272 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
