Assay Details
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205364.1 | NP_990695.1 | 2 - 3 | 411 | 80 | |
XM_025144691.1 | XP_025000459.1 | 1 - 2 | 101 | 80 | ||
GenBank mRNA | U48860.1 | - | 2 - 3 | 411 | 80 | |
X83889.1 | - | 270 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025144691.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
