Assay Details
Gene Symbol: GSTT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205365.1 | NP_990696.1 | 2 - 3 | 214 | 109 | |
GenBank mRNA | U13676.1 | - | 2 - 3 | 214 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
