Assay Details
Gene Symbol: GSTT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205365.1 | NP_990696.1 | 3 - 4 | 299 | 85 | |
GenBank mRNA | U13676.1 | - | 3 - 4 | 299 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
