Assay Details
Gene Symbol: LECT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205478.2 | NP_990809.2 | 5 - 6 | 656 | 72 | |
GenBank mRNA | CR388911.1 | - | 5 - 6 | 643 | 72 | |
M29449.1 | - | 5 - 6 | 656 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
