Assay Details
Gene Symbol: HMGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205486.1 | NP_990817.1 | 3 - 4 | 381 | 93 | |
XM_025149536.1 | XP_025005304.1 | 2 - 3 | 324 | 93 | ||
GenBank mRNA | BX935920.2 | - | 3 - 4 | 390 | 93 | |
M80574.1 | - | 3 - 4 | 398 | 93 | ||
M83235.1 | - | 3 - 4 | 381 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
