Assay Details
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001778.2 | NP_001001778.2 | 4 - 5 | 522 | 96 | |
GenBank mRNA | AB106870.1 | - | 4 - 5 | 520 | 96 | |
AJ720911.1 | - | 4 - 5 | 522 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
