Assay Details
Gene Symbol: APOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011692.1 | NP_001011692.1 | 4 - 5 | 406 | 98 | |
GenBank mRNA | AY735998.1 | - | 4 - 5 | 406 | 98 | |
BX934317.2 | - | 4 - 5 | 407 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
