Assay Details
Gene Symbol: EPGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012404.1 | NP_001012404.1 | 3 - 4 | 839 | 71 | |
XM_015276322.2 | XP_015131808.1 | 3 - 4 | 3645 | 71 | ||
GenBank mRNA | AY885230.1 | - | 3 - 4 | 839 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
