Assay Details
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012551.1 | NP_001012569.1 | 2 - 3 | 296 | 88 | |
GenBank mRNA | AJ851582.1 | - | 2 - 3 | 296 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
