Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012582.1 | NP_001012600.1 | 4 - 5 | 518 | 63 | |
GenBank mRNA | AJ851746.1 | - | 4 - 5 | 518 | 63 | |
CR523601.1 | - | 4 - 5 | 499 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
