Assay Details
Gene Symbol: FBXL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012786.1 | NP_001012804.1 | 2 - 3 | 267 | 88 | |
GenBank mRNA | AJ851502.1 | - | 2 - 3 | 267 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
