Assay Details
Gene Symbol: MOV10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012843.1 | NP_001012861.1 | 203 | 71 | ||
XM_015298929.2 | XP_015154415.1 | 348 | 71 | |||
GenBank mRNA | AJ720147.1 | - | 203 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
